IV Inspection Light Box, Healthcare Logistics
MSPP-17109
- Description:IV Inspection Light Box
- Dimensions:193/4×241/2×7"
- Cat. No.:MSPP-17109
This light box is used to determine the absence or inclusion of particulates in compounded sterile drug products and perform finished preparation release checks for purity and quality.
- Both lights are independently operated
- Predrilled mounting holes
- Flexible arm permits easy movement
Physical inspection of the compounded sterile product (CSP) is achieved against lighted White and Black backgrounds. 2× magnifying glass has Ø value of 4" is used for close visual inspection.
: One 110 V wall outlet is needed for operation. Supplied with LED lights (one black and one white), two 5' cords and two independent light switches.