Order Entry
Puerto Rico
ContactUsLinkComponent
Test Tube Support System, PP, Cole Parmer
Test Tube Support System, PP, Cole Parmer
Catalog # MSPP-EW-067330
Supplier:  Cole Parmer
undefined
Test Tube Support System, PP, Cole Parmer
Catalog # MSPP-EW-067330
Supplier:  Cole Parmer
Supplier Number:  06733-00

Specifications

  • Width:
    4"
  • Description:
    Test Tube Rack System
  • Length:
    9"
  • Material:
    PP
  • Cat. No.:
    MSPP-EW-067330

Specifications

About this item

Rotate each cube independently-use several sizes of test tubes, culture tubes, or microcentrifuge tubes at the same time.

  • Cube modules are color coded for convenience
  • Autoclavable

Compatible with the tubes: 15 to 50 ml conical; 12×75 mm, 13×100 mm, 16×125 mm, and 17×100 mm culture; and 1.5 and 2.0 ml microcentrifuge.

Delivery: Each system consists of four cubes and one support tray.