PELCO® Barrier Foil Resealable Storage Pouches, Ted Pella
Catalog # MSPP-139313
Supplier: TED PELLA INC MS
Specifications
- Material:Aluminium
- Environmentally Preferable:
- Cat. No.:MSPP-139313
- W×D×H:3¹/₂×10¹/₂×8"
Specifications
About this item
This barrier bag is ideal for storage of materials sensitive to moisture and/or oxygen. The inside dimensions measures with a resealable lock and a tear notch above the zipper.
- Heat sealable, and resealable
- Compatible with vacuum sealers
The material is aluminum foil covered multi-layered polyethylene, providing an MVTR (moisture vapor transmission rate) that is 1000 times better than a typical poly bag. Storage of light, moisture and oxygen sensitive materials/samples, Immunofluorescence labeled, Confocal or Immunology ANA slides from room temperature to freezer, radioactive labeled electrophoresis gels for both refrigerator and freezer and storage and transport of gas monitor badges.