Order Entry
Puerto Rico
ContactUsLinkComponent
Temp-Chex Digital II Laboratory Thermometer, Streck
Temp-Chex Digital II Laboratory Thermometer, Streck
Catalog # 10783-088
undefined
Temp-Chex Digital II Laboratory Thermometer, Streck
Catalog # 10783-088
Supplier Number:  240501

Specifications

  • Description:
    Temp-Chex digital II laboratory thermometer with bottle probe (filled with glass beads)
  • Cat. No.:
    10783-088
  • Accuracy:
    ±1 °C (−50...−30 °C), ±0.5 °C (−30...+90 °C), ±2 °C (90...+150 °C); ±3 °C (150...+200 °C)
  • Range:
    –50...+200 °C
  • Supplier no.:
    240501

Specifications

About this item

Temp-Chex digital II is a digital thermometer for monitoring refrigerators, freezers, incubators and room temperatures.

  • Easy-to-read display with minimum/maximum temperature, low/high alarm and date/time modes
  • Can be mounted magnetically, adhesively, by keyhole or flip-out desk stand
  • Thermometer probe is inserted in a vial of glass beads to buffer temperature variations
  • Automatically records the date and time of up to 10 events when a temperature is out of alarm limits

A free wall chart for daily temperature recording is included with each Temp-Chex Digital II. The thermometer probe is immersed in a vial of glass beads, so readings will not fluctuate with the opening and closing of doors, resulting in a more accurate reading.

Certifications: Certified NIST-traceable.