Dehydrated Media, Soyabean Casein Digest Medium (Tryptone Soya Broth), HiMedia Laboratories
Supplier: HiMedia
Certificates
About this item
Soyabean casein digest medium, granulated is a general purpose medium used for cultivation of a wide variety of microorganisms and recommended for sterility testing of moulds and lower bacteria.
- Cream to yellow coloured granular medium
- pH of 3.0% weight/volume aqueous solution at 25 °C
Store below 30 °C in a tightly closed container and the prepared medium at 2 to 8 °C.