Pluronic® F-68 Polyol
Catalog # IC092750016
Supplier: MP Biomedicals
CAS Number:
Specifications
- Size:100 g
- Cat. No.:IC092750016
Specifications
Specification Test Results
- Identity Test :Passes
Specification Test Results
About this item
:
:8400 g/mol
Melting Pt:52 °C (lit.)
Storage Temperature:Ambient
CAS Number:9003-11-6
MDL Number:MFCD00082049