VOYAGER II Adjustable Tip Spacing Pipetting Systems, INTEGRA Biosciences AG
Supplier: INTEGRA MS
Certificates
About this item
VOYAGER II pipettes feature with motorized tip spacing which helps in enabling parallel transfer of multiple samples between labware of different sizes and formats.
- GripTips snap into place with minimal tip loading effort, providing a secure connection
- Pipettes can be charged directly with the pipette mains adapter
- Versatile user interface and ergonomic design
- Dispensing multiple aliquots of the same volume
VOYAGER II pipettes feature with motorized tip spacing which helps in enabling parallel transfer of multiple samples between labware of different sizes and formats.
These pipettes provide access to a wide range of labware formats such as microplates, tube racks or gel chambers. These are ideal for a wide variety of genomic, proteomic and cell culture applications.
Ordering information: Power supplies are not included, need to order separately.