- Source:
- Species:
- Description:Mebiol® Thermoreversible Hydrogel
- Size:10 ml
- Environmentally Preferable:
- Cat. No.:103701-490
Advanced BioMatrix products are recognized for purity, functionality, quality, and lot-to-lot consistency, allowing for reproducibility in research.
- Unique sol-gel temperature dependent reversibility
- Biologically inert material with high transparency
- Great for stem cell culture, expansion, differentiation, and spheroid culture
Mebiol® thermoreversible hydrogel is a copolymer of poly(N-isopropylacrylamide) and poly (ethylene glycol) (PNIPAAm-PEG). The defining feature of Mebiol® is its temperature reversible sol-gel transition. Chill Mebiol® to turn it into a sol (meaning it handles like a liquid). Heat Mebiol® to turn it into a solid hydrogel. The unique thermoreversible feature allows for extremely easy cell handling. Cultures are seeded into cooled Mebiol® and recovered conveniently by cooling the culture vessel and centrifugation. In the gel state, the highly lipophilic and network structure of the Mebiol® presents an efficient niche for cell proliferation, cell communication, gas and mass exchange, and protection of cells and tissue from shear forces
Mebiol® is easy to use, non-toxic, 100% synthetic and has high transparency. Published applications include stem cell and pluripotent stem cell culture, expansion and differentiation, spheroid culture, cell implantation, organ and tissue regeneration, drug delivery and more. Though Mebiol® suppresses fibroblast growth, it has been commercialized as a cell/tissue culture reagent for ES cells, chondrocytes and cancer cells.