ContactUsLinkComponent
LSM™ Lymphocyte Separation Medium, MP Biomedicals
LSM™ Lymphocyte Separation Medium, MP Biomedicals
Catalog # 76202-438
Supplier:  MP Biomedicals
undefined
LSM™ Lymphocyte Separation Medium, MP Biomedicals
Catalog # 76202-438
Supplier:  MP Biomedicals
Supplier Number:  0850494X
Specifications
  • Cat. No.:
    76202-438
Specifications
About this item
LSM™ is a sterile-filtered solution of a sucrose polymer and diatrizoate salts.
  • LSM™ is formulated for isolation of mononuclear cells from defibrinated or heparinized whole human blood.
  • One-step centrifugation permits separation of mononuclear lymphocytes from erythrocytes, polynuclear lymphocytes and most platelets.