- Species:Human
- Size:50 ml
- Storage Conditions:Store at 2-8 °C.
- Protein/Peptide Name:Albumin
- Concentration:30.0 g/dL; 30% Solution
- Cat. No.:IC08823051
Albumin may be used to eliminate background interference in ELISA's or other enzyme assay systems. It is also used in absorption, distribution, metabolism, and excretion (ADME) pharmacological research; cell culture; drug delivery research; and cryopreservation of cells. Human and bovine albumins contain 16% nitrogen and are often used as standards in protein calibration studies. Due to their free hydrophobic region fatty acid free albumins are used to solubilize lipids in tissue culture, and are also used as blocking agents in Western blots or ELISA applications. Globulin free albumins are suitable for use in applications where no other proteins should be present (e.g., electrophoresis).
The source material from which this product is manufactured has been tested for the presence of Hepatitis B Surface Antigen (HBsAg), HIV I/II, HIV-1Ag, HCV, ALT and Syphilis. Product tested negative by currently approved FDA tests.
Human serum albumin undergoes three different post-translational modifications: oxidation, glycation, and S-nitrosylation. Modifications usually occur on the surface of the globular protein, and do not significantly affect conformation. However, modification strongly affects binding of fatty acids and drug molecules.