Formic acid 98-100% (by alkalimetry), EMPROVE® ESSENTIAL DAC, SAFC®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:64-18-6
:
Formula:HCO₂H
MDL Number:MFCD00003297
:46.03 g/mol
Boiling Pt:101 °C (1013 hPa)
Melting Pt:4 °C
Density:1.22 g/cm³ (20 °C)
Flash Pt:49.5 °C
Storage Temperature:Ambient
EINECS:200-579-1
UN:1779
ADR:8 (3),II
:GHS02
:GHS05
:GHS06
Specification Test Results
- Assay (Alkalimetric):≥98.0%
- Identity (IR-Spectrum):Passes Test
- Density (d 20/20):1.217 - 1.223
- Chromatographic Purity (Acetic acid):≤0.5%
- Chromatographic Purity (Other Impurity):≤0.1%
- Chromatographic Purity (Sum of All Other Impurities):≤0.3%
- Chloride (Cl):≤0.001%
- Sulfate (SO₄):≤0.004%
- Sulfite (SO₃):≤0.002%
- Heavy Metals (as Pb):≤0.0005%
- As (Arsenic):≤0.0003%
- Cu (Copper):≤0.002%
- Pb (Lead):≤0.001%
- Zn (Zinc):≤0.002%
- Aldehydes:Passes Test
- Formaldehyde (HCHO):≤0.1%
- Oxalic Acid (C₂H₂O₄):≤0.01%
- Residual Solvents (ICHQ3C) of Class 2 (Methanol):≤0.3%
- Evaporation Residue:≤0.005%
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_1.00263
SDS
VWR GHS Compliance Letter (US)