Order Entry
Puerto Rico
ContactUsLinkComponent
CytoSoft® Discovery Kit, Tunable Substrate Stiffness,Of Each Stiffness, Advanced BioMatrix
CytoSoft® Discovery Kit, Tunable Substrate Stiffness,Of Each Stiffness, Advanced BioMatrix
Catalog # 103701-726
Supplier:  Advanced Biomatrix
undefined
CytoSoft® Discovery Kit, Tunable Substrate Stiffness,Of Each Stiffness, Advanced BioMatrix
Catalog # 103701-726
Supplier:  Advanced Biomatrix
Supplier Number:  5190-7EA

Specifications

  • Description:
    Cytosoft® Discovery Kit, Tunable Substrate Stiffness, Of Each Stiffness
  • Cat. No.:
    103701-726

Specifications

About this item

Advanced BioMatrix products are recognized for purity, functionality, quality, and lot-to-lot consistency, allowing for reproducibility in research.

  • Substrate of specific stiffness
  • Each lot is measured and validated
  • Full physiological range available
  • Test the entire range (0.2 to 64kPa) with the discovery kit

This CytoSoft® product, Catalog Number 5190-7EA, has various elastic moduli of approximately 0.2, 0.5, 2, 8, 16, 32 and 64 kPa in seven individual 6-well plates. The thickness of the silicone gel is uniform with a ~0.5 mm thick layer of silicone in each dish that is fully compatible with mammalian cell cultures. The plates are packaged individually, sterilized using irradiation.

The rigidity of the substrate to which cells adhere can have a profound effect on cell morphology and gene expression. CytoSoft® products provide a tool to culture cells on substrates with various rigidities covering a broad physiological range. On the bottom of each well, there is a thin layer of specially formulated biocompatible silicone, whose elastic modulus (rigidity) is carefully measured and certified. The surfaces of the gels in CytoSoft® products are functionalized to form covalent bonds with amines on proteins. This chemical functionalization is stable, and the reaction does not require a catalyst, facilitating the coating of the gel surfaces with matrix proteins and plating cells.

The silicone substrates of CytoSoft® products are optically clear and have a low auto-florescence. The layer of silicone in each dish is firmly bonded to the bottom of the dish. Unlike hydrogels (such as polyacrylamide gels), silicone gels are not susceptible to hydrolysis, do not dry nor swell, are resilient and resistant to tearing or cracking, and their elastic moduli (rigidities) remain nearly unchanged during extended storage periods.

CytoSoft® products accommodate the harvesting of cells using enzymes such as trypsin and collagenase. There is no biochemical breakdown of the substrate during or after enzyme treatment, and there are no residuals of the substrate in the sample retrieved from a CytoSoft® plate.

Caution: This product is for R and D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.