ContactUsLinkComponent
PureCol® Type I Collagen Coated Flasks, Advanced BioMatrix
PureCol® Type I Collagen Coated Flasks, Advanced BioMatrix
Catalog # 103700-650
undefined
PureCol® Type I Collagen Coated Flasks, Advanced BioMatrix
Catalog # 103700-650
Supplier Number:  5029-10EA
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Purecol® Type I Collagen Coated Flasks
  • Environmentally Preferable:
  • Cat. No.:
    103700-650
Specifications
About this item
Advanced BioMatrix products are recognized for purity, functionality, quality and lot-to-lot consistency, allowing for reproducibility in research.
  • Coated with PureCol® - the collagen standard for purity
  • Great for 2D cell culture on a native ECM substrate
  • PureCol® is referenced in >2000 publications
PureCol® Collagen Coated, T-25 flasks with vented caps has a uniform and consistent application of high-quality Type I collagen on clear polystyrene surface. Flasks have an anti-drip edge and modified canted neck to provide maximum control replacing nutrient depleted media. The flasks have an anti-tip feature and specially shaped pouring channel for improved handling. Flasks are aseptically processed and are non-pyrogenic.
Caution: This product is for R and D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.