Order Entry
Puerto Rico
ContactUsLinkComponent
10X Phosphate Buffered Saline (PBS), Sterile, Dulbecco's Formula, without Magnesium and Calcium, MP Biomedicals
10X Phosphate Buffered Saline (PBS), Sterile, Dulbecco's Formula, without Magnesium and Calcium, MP Biomedicals
Catalog # IC091960454
Supplier:  MP Biomedicals
CAS Number:  
10X Phosphate Buffered Saline (PBS), Sterile, Dulbecco's Formula, without Magnesium and Calcium, MP Biomedicals
Catalog # IC091960454
Supplier:  MP Biomedicals
Supplier Number:  091960454
CAS Number:  

Specifications

  • Description:
    10X Phosphate buffered saline (PBS), sterile, Dulbecco's formula
  • Size:
    500 ml
  • Storage Temperature:
    15 to 30° C
  • Cat. No.:
    IC091960454
  • pH:
    7.0 - 7.6

Specifications

About this item

Role of a balanced salt solution in cell culture is multifaceted and can be divided into four principal functions: 1. Serves as an irrigating, transporting and diluting fluid while maintaining intra- and extracellular osmotic balance; 2. Provides cells with water and certain bulk inorganic ions essential for normal cell metabolism; 3. Combined with a carbohydrate, such as glucose, provides the principal energy source for cell metabolism; 4. Provides a buffering system to maintain the medium within the physiological pH range (7.2 - 7.6)

  • Phosphate Buffered Salts, Dulbecco's Formula (DPBS) is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells.
  • DPBS is used to to irrigate, wash, and dilute mammalian cells.
  • Phosphate buffering maintains the pH in the physiological range.
  • Calcium and magnesium facilitate cell binding and clumping.
  • DPBS without these ions can be used to wash and rinse suspended cells.

Inorganic Salts: Potassium Chloride [KCl]: 200.00 mg/L; Potassium Phosphate Monobasic [KH2PO4]: 200.00 mg/L; Sodium Chloride [NaCl]: 8000.00 mg/L; Sodium Phosphate Dibasic [Na2HPO4]: 1150.00 mg/L

Sterile
Osmolality: 270 - 305 mM/kg
Clear colorless solution