Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium pyruvate 11.00mg/ml 100 mM, sterile
Sodium pyruvate 11.00mg/ml 100 mM, sterile
Catalog # IC091682049
Supplier:  MP Biomedicals
CAS Number:  
undefined
Sodium pyruvate 11.00mg/ml 100 mM, sterile
Catalog # IC091682049
Supplier:  MP Biomedicals
Supplier Number:  091682049
CAS Number:  

Specifications

  • Size:
    100 ml
  • Cat. No.:
    IC091682049

Specifications

About this item

 :
 :110.04 g/mol
Density:~1.267 g/mL (lit.)
Storage Temperature:Refrigerator
CAS Number:113-24-6
Formula:CH₃COCOONa
MDL Number:MFCD00002586