ContactUsLinkComponent
LaMotte® TesTab® Dissolved Oxygen/BOD Kit
LaMotte® TesTab® Dissolved Oxygen/BOD Kit
Catalog # 470326-578
Supplier:  LaMotte
CAS Number:  
undefined
LaMotte® TesTab® Dissolved Oxygen/BOD Kit
Catalog # 470326-578
Supplier:  LaMotte
Supplier Number:  5889
CAS Number:  
Specifications
  • Description:
    Dissolved Oxygen/BOD Test Kit
  • Size:
    1 Kit
  • Cat. No.:
    470326-578
  • No. of tests:
    100
Specifications
About this item
Refill for The Comprehensive Water Monitoring Kit 470004-240 and LaMotte® TesTab® Water Investigation Kit 470007-270
  • 100 Tests
  • Range 0,4,8 ppm
Each kit contains easy-to-read diagrammed instructions, a color chart, a test factor flash card and all the necessary apparatus and non-hazardous TesTabs® to perform 100 tests