Pipette Stand, Rotary, Pipette Carousel 2, Eppendorf
These pipette carousels holds up to six manual single or multi channel pipettes.
- Large carrying handle for easy transport of carousel from bench to bench
- Large rubber feet protect stand from liquids spilled on lab bench
- Small footprint with a diameter of 18.5 cm
- Single pipette holders can be mounted to the carousel in two orientations turned by 180° to each other
- Interchangeable pipette holders, no tools required
- Pictograms clearly label pipette holders for easy assignment in the lab
These are without charging functionality.
: Sticky tape included. Additional pipette holders are available separately.
- Catalog No:
- 76204-692
- 76204-694
- 76204-696
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001