PureCol® Type I Collagen Coated Dishes, Advanced BioMatrix
Catalog # 103700-648
Supplier: Advanced Biomatrix
Specifications
- Ø:100 mm
- Description:Purecol® Type I Collagen Coated Dishes
- Height:20 mm
- Cat. No.:103700-648
Specifications
About this item
Advanced BioMatrix products are recognized for purity, functionality, quality, and lot-to-lot consistency, allowing for reproducibility in research.
- Coated with PureCol® - the collagen standard for purity
- Great for 2D cell culture on a native ECM substrate
- PureCol® is referenced in >2000 publications
PureCol® collagen coated dishes have a uniform and consistent application of high quality type I collagen on clear polystyrene surface. Plates have a raised ring around the circumference (SureGrip) of the dish that provides a secure grip of both the dish and lid. Embossed arrows on the dish and lid provide orientation for cell site observation. Dishes are aseptically processed and are non-pyrogenic.
Caution: This product is for R and D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.