Order Entry
Puerto Rico
ContactUsLinkComponent
Rat C-Peptide ELISA Kit, ALPCO
Rat C-Peptide ELISA Kit, ALPCO
Catalog # 101720-806
Supplier:  Alpco Diagnostics
CAS Number:  
Rat C-Peptide ELISA Kit, ALPCO
Catalog # 101720-806
Supplier:  Alpco Diagnostics
Supplier Number:  80-CPTRT-E01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Rat C-Peptide ELISA
  • Sample Type:
    Serum
  • Detection Range:
    50 - 4500 pM
  • Sensitivity:
    10.8 pM
  • Cat. No.:
    101720-806
  • Supplier no.:
    80-CPTRT-E01
  • No. of tests:
    96 wells

Specifications

About this item

The Rat C-peptide ELISA quantifies the C-peptide protein products of rat I and rat II proinsulin genes.

  • 10 µl of sample is sufficient for the assay
Caution: For research use only. Not for use in diagnostic procedures.