Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Dextrose solution 5% USP for injection

Plastic container, Viaflex®, 250 ml
Catalog # 68000-132
Supplier:   Baxter
 :  2B0062Q
Dextrose solution 5% USP for injection
Catalog # 68000-132
Supplier:  Baxter
CAS Number:  50-99-7
Dextrose solution 5% USP for injection
Dextrose solution 5% USP for injection

Plastic container, Viaflex®, 250 ml

CAS Number:  50-99-7
Price
$48.87
Log in for availability
Each(1/Itm/Bx) Case of 36 (1/ Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
  • Total calories: 170 kcal/l
  • Specific gravity: 1.02
  • Fill range volume (ml): 265 to 285
  • Store at room temperature (25 °C)
  • pH : 4.0 (3.2 - 6.5)
  • mOsmol/L (calc): 252
  • NDC Number: 0338-0017-02

 : USP specified.

Synonyms:  Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
  • CAS Number:50-99-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148912
  • Molecular Weight:180.16 g/mol

 

Specifications

Specifications

  • Pack type:
    Plastic container, Viaflex®
  • Size:
    250 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.