To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Bottle, amber, butyl rubber stopper
- Source:
- Species:
- Size:1.1 M NP units
- Environmentally Preferable:
- Storage Conditions:<–20 °C
- Form:Solid
- Enzyme Name:BP Protease
- Cat. No.:101449-128
- Supplier no.:003-1000
Specifications
About this item
BP Protease is an affordable, aseptically dispensed, lyophilized protein containing purified neutral protease from Bacillus polymyxa.
- Low contaminating endotoxin levels (<50 EU/OD)
- Improved performance
- Vial contains 1.1 million neutral protease (NP) units, with total NP activity >800000 NP units
BP Protease is a Dispase™ equivalent, neutral protease enzyme, a superb product to isolate human epithelial skin cells from tissue or after in vitro culture. This enzyme is also a component of the DE collagenase products and is used in enzyme mixtures that also contain collagenase HA and collagenase MA.
These combinations have been successfully used to isolate islets, hepatocytes, and adipose derived stromal cells (ADSC), and are versatile and flexible enough to use for many other cell isolation applications.