Human Recombinant SIRP alpha (from HEK293 Cells), Fc Tag
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein Function:Immune Checkpoint Proteins
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CD47, His Tag (Cat. No. CD7-H5227) at 2 μg/mL can bind Human SIRP alpha, mouse IgG1 Fc Tag (HPLC-verified) (Cat. No. SIA-H52A8) with a linear range of 8-250 ng/mL (QC tested).; FACS assay shows that recombinant Human SIRP alpha, mouse IgG1 Fc Tag (HPLC-verified) (Cat. No. SIA-H52A8) can bind to Jurkat cell expressing CD47. The concentration of SIRP alpha used is 0.3 μg/ml (Routinely tested).; FACS analysis shows that the binding of Human SIRP alpha, mouse IgG1 Fc Tag (HPLC-verified) (Cat. No. SIA-H52A8) to Jurkat expressing CD47 was inhibited by increasing concentration of neutralizing anti-CD47 antibody. The concentration of SIRP alpha used is 0.3 μg/ml. IC50=0.1318 μg/ml (Routinely tested).
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:MYD1|PTPNS1|CD172A|SHPS1|P84|SIRPA|BIT|MFR
- UniProtKB:P78324
- Protein/Peptide Name:SIRP alpha
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:63.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:Powder,RT