Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Human recombinant S100A8 (from E. coli)

Marketsource
Supplier:   ACRO Biosystems
Human recombinant S100A8 (from <i>E. coli</i>)
Supplier:  ACRO Biosystems
Marketsource
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 103015-356
  • 103015-354
  • Protein/Peptide Name:
  • S100A8
  • S100A8
  • Protein Synonyms:
  • CP-10|S100A8|CFAG|CAGA|MRP8|NIF|CGLA|60B8AG|P8
  • CP-10|S100A8|CFAG|CAGA|MRP8|NIF|CGLA|60B8AG|P8
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Protein Function:
  • Bio-Markers & CD Antigens
  • Bio-Markers & CD Antigens
  • Species:
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Protease-free:
  • Y
  • Y
  • Animal-Free:
  • Y
  • Y
  • Endotoxin Level:
  • Low
  • Low
  • Endotoxin Content:
  • Less than 1.0 EU per μg by the LAL method.
  • Less than 1.0 EU per μg by the LAL method.
  • UniProtKB:
  • P05109
  • P05109
  • Biological Activity:
  • Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Purity:
  • >95% as determined by SDS-PAGE.
  • >95% as determined by SDS-PAGE.
  • Formulation:
  • Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Molecular Weight:
  • 10.8 kDa
  • 10.8 kDa
  • Storage Conditions:
  • –20 °C
  • –20 °C
  • Shipping Temperature:
  • Powder,RT
  • Powder,RT

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100 μg
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $282.00
    $282.00
    Each (100µG)
       $282.00

    Supplier Number: S18H5112100UG
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    103015-356
    Protein/Peptide Name
    S100A8
    Protein Synonyms
    CP-10|S100A8|CFAG|CAGA|MRP8|NIF|CGLA|60B8AG|P8
    Protein/Peptide Type
    Recombinant
    Protein Function
    Bio-Markers & CD Antigens
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Protease-free
    Y
    Animal-Free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    Less than 1.0 EU per μg by the LAL method.
    UniProtKB
    P05109
    Biological Activity
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
    Purity
    >95% as determined by SDS-PAGE.
    Formulation
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
    Molecular Weight
    10.8 kDa
    Storage Conditions
    –20 °C
    Shipping Temperature
    Powder,RT
    Marketsource
  • 1 mg
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $1,740.00
    $1,740.00
    Each (1mg)
       $1,740.00

    Supplier Number: S18H51121MG
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    103015-354
    Protein/Peptide Name
    S100A8
    Protein Synonyms
    CP-10|S100A8|CFAG|CAGA|MRP8|NIF|CGLA|60B8AG|P8
    Protein/Peptide Type
    Recombinant
    Protein Function
    Bio-Markers & CD Antigens
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Protease-free
    Y
    Animal-Free
    Y
    Endotoxin Level
    Low
    Endotoxin Content
    Less than 1.0 EU per μg by the LAL method.
    UniProtKB
    P05109
    Biological Activity
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
    Purity
    >95% as determined by SDS-PAGE.
    Formulation
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
    Molecular Weight
    10.8 kDa
    Storage Conditions
    –20 °C
    Shipping Temperature
    Powder,RT
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.