Order Entry
Puerto Rico
ContactUsLinkComponent
 
Recombinant Protein L (from E. coli)
 
Recombinant Protein L (from E. coli)

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in 100 mM NaH2PO4, pH8.5. Normally trehalose is added as protectant before lyophilization.
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    RPL|Protein L
  • Protein/Peptide Name:
    Protein L
  • Purity:
    >95% as determined by SDS-PAGE.
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 100 mM NaH2PO4, pH8.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    Powder,RT