Human Recombinant PD-L1 (from HEK293 Cells), Fc Tag
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein Function:Immune Checkpoint Proteins
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human PD-1, Strep Tag (Cat. No. PD1-H5284) at 2 μg/mL (100 μL/well) can bind Human PD-L1, Fc Tag (HPLC-verified) (Cat. No. PD1-H5258) with a linear range of 4-63 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human PD-L1, Fc Tag (HPLC-verified) (Cat. No. PD1-H5258) at 5 μg/mL (100 μL/well) can bind Anti-PD-L1 MAb with a linear range of 0.2-2 ng/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized Human B7-1, mouse IgG2a Fc Tag, low endotoxin (Cat. No. B71-H52A4) at 5 μg/mL (100 μL/well) can bind Human PD-L1, Fc Tag (HPLC-verified) (Cat. No. PD1-H5258) with a linear range of 0.01-0.313 μg/mL (Routinely tested).; Flow Cytometry assay shows that recombinant Human PD-L1, Fc Tag (HPLC-verified) (Cat. No. PD1-H5258) can bind to 293 cell overexpressing human PD-1. The concentration of PD-L1 used is 10 μg/mL (Routinely tested).; FACS analysis shows that the binding of Human PD-L1, Fc Tag (HPLC-verified) to 293 overexpressing PD-1 was inhibited by increasing concentration of neutralizing Anti-PD-L1 antibody. The concentration of PD-L1 used is 10 μg/mL. The IC50 is 12.92 μg/mL (Routinely tested).; Immobilized cell surface PD-1 (5x104 of cells per well) can bind Human PD-L1, Fc Tag (HPLC-verified) (Cat. No. PD1-H5258) with an EC50 of 0.029 μg/mL (Routinely tested).
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
- UniProtKB:Q9NZQ7
- Protein/Peptide Name:PD-L1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:51.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:Powder,RT