Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant PD-L1 (from HEK293 Cells) His Tag
 
Human Recombinant PD-L1 (from HEK293 Cells) His Tag

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immune Checkpoint Proteins
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human PD-L1, His Tag (HPLC-verified) (Cat. No. PD1-H5229) at 1 μg/mL (100 μL/well) can bind Anti-Human PD-L1 MAb (Human IgG1) with a linear range of 0.1-3 ng/mL (QC tested).; Biotinylated Human PD-1, Fc Tag (Cat. No. PD1-H82F1) at 1 μg/mL (5 μL/well) can bind Human PD-L1, His Tag (HPLC-verified) (Cat. No. PD1-H5229) with a linear range of 0.02-0.625 μg/mL as determined in a alphaLISA homogeneous assay (Routinely tested).; Human PD-1, Fc Tag (Cat. No. PD1-H5257) captured on CM5 chip via anti-human IgG Fc antibody, can bind Human PD-L1, His Tag (HPLC-verified) (Cat. No. PD1-H5229) with an affinity constant of 3.6 μM as determined in a SPR assay (Biacore T200) (QC tested).; Anti-Human PD-L1 Mab (Human IgG1) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Human PD-L1, His Tag (Cat. No. PD1-H5229) with an affinity constant of 0.286 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
  • UniProtKB:
    Q9NZQ7
  • Protein/Peptide Name:
    PD-L1
  • Purity:
    >98% as determined by SDS-PAGE.
  • Molecular Weight:
    26.0 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    Powder,RT