Human Recombinant IL-2 R beta (from HEK293 cells)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein Function:Bio-Markers & CD Antigens
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.; Human IL-2 R beta Protein, Fc Tag (Cat. No. ILB-H5253) captured on CM5 chip via anti-human IgG Fc antibody, can bind IL-2 Protein, Tag Free (Cat. No. IL2-H4113) with an affinity constant of 0.6 μM as determined in a SPR assay (Biacore T200) (QC tested).; Human IL-2 R beta Protein, Fc Tag (Cat. No. ILB-H5253) captured on CM5 chip via anti-human IgG Fc antibody, can bind Human IL-15 Protein, Tag Free (Cat. No. IL5-H4217) with an affinity constant of 9.5 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD122|P70-75|IL2RB|RP5-1170K4.6
- UniProtKB:P14784
- Protein/Peptide Name:IL-2 R beta
- Purity:>92% as determined by SDS-PAGE.
- Molecular Weight:50.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:Powder,RT