Order Entry
Puerto Rico
ContactUsLinkComponent
 
Avitag™ Cynomolgus macaque;Rhesus macaque Recombinant CTLA-4 (from HEK293 cells), Acro Biosystems
Catalog # 103014-272
Supplier:  ACRO Biosystems
CAS Number:  
Avitag™ Cynomolgus macaque;Rhesus macaque Recombinant CTLA-4 (from HEK293 cells), Acro Biosystems
Catalog # 103014-272
Supplier:  ACRO Biosystems
Supplier Number:  CT4C82E5200UG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
    Rhesus monkey
  • Size:
    200 μg
  • Tag sequence:
    Avi Tag (Avitag™)
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 2 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avi Tag, His Tag (Cat. No. CT4-C82E5) with a linear range of 2-31 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human B7-2, Fc Tag (Cat. No. CD6-H5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avi Tag, His Tag (Cat. No. CT4-C82E5) with a linear range of 2-63 ng/mL (Routinely tested).
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    CTLA4|CD152
  • UniProtKB:
    G7PL88
  • Protein/Peptide Name:
    CTLA-4
  • Purity:
    >92% as determined by SDS-PAGE.
  • Molecular Weight:
    15.9 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    Powder,RT
  • Cat. No.:
    103014-272
  • Supplier no.:
    CT4C82E5200UG

Specifications

About this item