Order Entry
Puerto Rico
ContactUsLinkComponent
 
Ebolavirus Recombinant Glycoprotein / GP (virus) (from HEK293 cells)
 
Ebolavirus Recombinant Glycoprotein / GP (virus) (from HEK293 cells)

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Ebolavirus
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Glycoprotein GP55|Envelope glycoprotein B
  • UniProtKB:
    B8XCN1
  • Protein/Peptide Name:
    Glycoprotein / GP (virus)
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    34.3 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    Powder,RT

 

Frequently Bought Together