Eosin B alcoholic solution Optik Type 2
500 ml
Catalog # 76270-178
Supplier: Avantik
Supplier #: RS4365-A
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Optik Type 2 Eosin is an alcoholic eosin with exceptional contrast between cytoplasmic components and nuclei.
- Stains cytoplasm various shades of pink and red.
Synonyms: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91
- CAS Number:548-24-3
- Formula:C₂₀H₆Br₂N₂Na₂O₉
- MDL Number:MFCD00005041
- Molecular Weight:624.06 g/mol
Specifications
Specifications
- Size:500 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



