Order Entry
Puerto Rico
ContactUsLinkComponent
FibriCol® Bovine Collagen Type I
FibriCol® Bovine Collagen Type I
Catalog # 103700-658
CAS Number:  
undefined
FibriCol® Bovine Collagen Type I
Catalog # 103700-658
Supplier Number:  5133-20ML
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Species:
    Bovine
  • Size:
    20 ml
  • Protein/Peptide Name:
    Collagen Type I
  • Cat. No.:
    103700-658

Specifications

About this item

FibriCol® collagen is known as the standard of all collagens for purity (>99% collagen content), functionality, and the most native-like collagen available. FibriCol® is isolated from bovine hides sourced from the only controlled, closed herd in the United States. Advanced BioMatrix’s manufacturing processes comply with stringent quality standards that have proven to yield unsurpassed lot-to-lot consistency.

FibriCol® collagen is approximately 97% Type I collagen with the remainder being comprised of Type III collagen. It contains a high monomer content as measured by gel permeation chromatography.

FibriCol® collagen is supplied at approximately a 10 mg/mL concentration. The concentration for each specific lot is provided on a Certificate of Analysis that is available with the purchase of each product. FibriCol® is soluble atelo-collagen in 0.01 N HCI, therefore, the pH is approximately 2.0.

FibriCol® collagen is ideal for applications where high concentrations of collagen are needed. FibriCol® collagen is provided in a 20 mL volume and is contained in user-friendly packaging for use and storage. FibriCol® is sterile filtered and is supplied as a ready to use solution.

  • Great for 2D Coatings
  • Highly purified collagen
  • Over 15 QC tests to ensure consistency and quality
Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.