Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human E-Cadherin

Human E-Cadherin

0.1 mg
Catalog # 103700-656
Marketsource
Supplier:   Advanced Biomatrix
Supplier #:  5085-0.1MG
Human E-Cadherin
Catalog # 103700-656
Supplier:  Advanced Biomatrix
Marketsource
CAS Number:  
Human E-Cadherin

0.1 mg

CAS Number:  
Price
$340.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

E-Cadherin has been used as coating matrix protein for 1) maintaining long-term ES or iPS cell culture then combine with proper ES cell culture media, 2) as a coating matrix material for 11R tag recombinant TF intracellular delivery for protein derived iPS protocol with extremely low-level non-specific interaction and 3) as a native antigen for antibody production.

  • Great for human ES cell culture
  • Sterility tested

Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.

Specifications

Specifications

  • Size:
    0.1 mg
  • Cat. No.:
    103700-656
  • Protein/Peptide Name:
    E-Cadherin
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.