Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Vitronectin

Human Vitronectin

0.1 mg
Catalog # 103701-198
Marketsource
Supplier:   Advanced Biomatrix
Supplier #:  5051-0.1MG
Human Vitronectin
Catalog # 103701-198
Supplier:  Advanced Biomatrix
Marketsource
CAS Number:  
Human Vitronectin
Human Vitronectin

0.1 mg

CAS Number:  
Price
$235.38
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Vitronectin is a monomeric glycoprotein used to promote cell attachment, migration, proliferation and differentiation in a broad number of cell lines and types. This product has been purified from human plasma where it is found as a mixture of 75kDa and 65kDa polypeptides.

Vitronectin’s primary use in cell culture is related to cell adhesion. It also binds to heparin and collagen.

Vitronectin is ideal for coating of surfaces. The optimal concentration for cell attachment and culture may differ for various cell types. Vitronectin has been used at a final coating concentration as low as 50 ng/cm2 on plasticware. It is provided in user-friendly packaging for use and storage. Vitronectin is sterile filtered and is supplied as a ready to use solution after thawing and concentration adjustment. This product is shipped separately on dry ice.

  • Great for 2D coatings
  • Highly purified, native ECM
  • Known for purity, consistency and quality
  • Purified from human plasma

Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.

Specifications

Specifications

  • Size:
    0.1 mg
  • Cat. No.:
    103701-198
  • Protein/Peptide Name:
    Vitronectin
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.