To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Size:50 ml
- Form:Solution
- Protein/Peptide Name:Poly-L-Lysine
- Molecular Weight:70,000 to 150,000 Da
- Concentration:0.1 mg/ mL
- Cat. No.:103701-192
Poly-L-Lysine is a synthetic amino acid chain that is positively charged having one hydrobromide per unit of Lysine. Poly-L-Lysine is widely used as a coating to enhance cell attachment and adhesion to both plasticware and glass surfaces. This molecule has been used to culture a wide variety of cell types. Certain cell types secrete proteases, which can digest Poly-L-Lysine. For those cell types, Poly-D-Lysine 000 Da) being less viscous and higher molecular weight (>300,000 Da) having more binding sites per molecule. This product’s molecular weight ranges from 70,000 to 150,000 Da yielding a solution viscosity for easy handling while providing sufficient binding sites for cell attachment.
Poly-L-Lysine surface coatings are designed to improve cell attachment, growth and differentiation of many cell types. Coated surfaces will often improve cell attachment in reduced or serum-free conditions. This product is supplied in a sterile 50 ml package size at a concentration of 0.1 mg/ml.
- Enhances cell attachment to plasticware and glass