To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Species:Bovine
- Size:1 Kit
- Protein/Peptide Name:Telocollagen Type I
- Concentration:6 mg/mL
- Cat. No.:103701-184
TeloCol®-6 bovine collagen solution is derived from an acid extraction process yielding a telopeptide-intact collagen. The pro-peptide regions at both ends of the collagen chain, N- and C-telopeptide regions, are maintained.
TeloCol®-6 collagen is approximately 95% Type I collagen with the remainder being comprised of Type III collagen. Each product includes a bottle containing 50 ml of collagen solution accompanied with a bottle of pre-formulated neutralizing solution for the formation of a collagen gel. This product is supplied at a concentration of approximately 6 mg/ml with the concentration for each specific lot provided on the product label and Certificate of Analysis that is available with the purchase of each product. TeloCol®-6 is sterile filtered and provided in user-friendly packaging for use and storage.
TeloCol®-6 is ideal for coating of surfaces, providing preparation of thin layers for culturing cells, or use as a solid gel.
- Great for 3D Hydrogels or 2D Coatings
- Highly purified collagen
- Over 15 QC tests to ensure consistency and quality
- Neutralization solution for easy 3D hydrogels