ContactUsLinkComponent
SUPERECORDER™ Portable/Wireless Universal Circular Chart, Graphic Controls
SUPERECORDER™ Portable/Wireless Universal Circular Chart, Graphic Controls
Catalog # 76240-364
Supplier:  Graphic Controls
SUPERECORDER™ Portable/Wireless Universal Circular Chart, Graphic Controls
Catalog # 76240-364
Supplier:  Graphic Controls
Supplier Number:  CT485-CWF
Specifications
  • Description:
    CTXL-TRH temperature/RH circular chart papers for 1 day, 100 charts
  • Cat. No.:
    76240-364
Specifications
About this item
The circular chart recorders from OMEGA combine form and function in an easy to use two-channel circular chart recorder with a large. It has a custom LCD readout that displays the current values of both channels, and can indicate the min, max or average input values as well.
  • High and low alarm indication
  • Keypad lockout status
It features a rugged, ABS enclosure, suitable for industrial environments, but its also intended to fit right in to other environments, such as museums, art galleries, computer rooms, labs, clean rooms, even hospitals. It is designed for both benchtop and wall mount operation; a convenient base rotates out of the way, and is hidden by a decorative foot cover for wall mount use.