Endosafe® 96-Well Plates, Charles River Endosafe
Catalog # MSPP-M9005
Supplier: CHARLES RIVER ENDOSAFE MS
Specifications
- Description:96-Well plates
- Material:Polystyrene
- Cat. No.:MSPP-M9005
- Supplier no.:M9005
Specifications
About this item
The plates are individually wrapped and tissue culture treated.
Certifications: Certified to 0.005 EU/ml