Order Entry
United States
ContactUsLinkComponent
Gas Detector Tubes for Carbon Monoxide, Sensidyne
Gas Detector Tubes for Carbon Monoxide, Sensidyne
Catalog # 14231-010
Supplier:  Sensidyne
Small Business Enterprise
undefined
Gas Detector Tubes for Carbon Monoxide, Sensidyne
Catalog # 14231-010
Supplier:  Sensidyne
Supplier Number:  106SA
Small Business Enterprise

Specifications

  • Temperature range:
    0...40 ºC
  • Cat. No.:
    14231-010
  • Measurement range:
    20 - 1000 ppm (100 ml), 5 - 50 ppm (400 ml)
  • No. of strokes:
    1 (100 ml) or 4 (400 ml)
  • Color change:
    Yellow to dark brown
  • Detectable gas:
    Carbon monoxide
  • Supplier no.:
    106SA

Specifications

About this item

Gas detector tubes are used for detection of airbourne gases and vapors.

  • Contains an antibacterial grip
  • Easy to use

These detector tubes safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors at a very low cost-per sample. These tubes are easy to use and provide a low-cost method of routine or spot sampling for gases and vapors.