Order Entry
United States
ContactUsLinkComponent
 
Gas Detector Tubes for Acetone, Sensidyne
  13502-652
 :  Sensidyne
Small Business Enterprise
undefined
Gas Detector Tubes for Acetone, Sensidyne
  13502-652
 :  Sensidyne
 :  102SA
Small Business Enterprise

 

  • Temperature range:
    0...40 ºC
  • Cat. No.:
    13502-652
  • Measurement range:
    1−5% (50 ml), 0.1-2.0% (100 ml)
  • No. of strokes:
    ½ (50 ml) or 1 (100 ml)
  • Color change:
    Orange to brown
  • Detectable gas:
    Acetone
  • Supplier no.:
    102SA

 

 

Gas detector tubes are used for detection of airbourne gases and vapors.

  • Contains an antibacterial grip
  • Easy-to-use

These detector tubes safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors at a very low cost-per sample. These tubes are easy to use and provide a low-cost method of routine or spot sampling for gases and vapors.