These mouse tail illuminator restrainers efficiently illuminates and warms a mouse tail for easy injection or bleeding.
- Restraining tube slides off base for easy cleaning
- Illuminated slot for easy tail vein injection
Features sliding doors to keep mouse gently restrained.