Grafco® Tincture Of Green Soap U.S.P.
Catalog # 82020-142
Supplier: GF HEALTH PRODUCTS INC. TE
Specifications
- Capacity:3800 ml (1 gal.)
- Description:Tincture of green soap
- Environmentally Preferable:
- Cat. No.:82020-142
- Supplier no.:3375
Specifications
About this item
Ideal for removal of dried blood and protein soils from skin, scalp, and hands.
- Distinctive oil of lavender scent
- Made in the USA
This vegetable-based alkaline detergent is made from pure vegetable oil, glycerin, and lavender oil. It is 30% alcohol by volume.