Masterflex® L/S® Precision PharmaPure Pump Tubings, Cole Parmer
: Avantor
Masterflex® L/S® PharmaPure pump tubings are used for filtration of sensitive and costly fluids.
- Ultra-low spallation protects fluid purity
- Low extractables and low gas permeability
- Long pumping life
- Pump at continuous pressures up to 40 psi
Nontoxic and nonhemolytic with excellent biocompatibility.
: Meets USP Class VI criteria.