ContactUsLinkComponent
Adapter, PTFE, Screw Thread, Analytical Sales and Services
Adapter, PTFE, Screw Thread, Analytical Sales and Services
Catalog # MSPP-CA4505
Adapter, PTFE, Screw Thread, Analytical Sales and Services
Catalog # MSPP-CA4505
Supplier Number:  CA4505
Specifications
  • Description:
    Thread adapter
  • Material:
    PTFE
  • Cat. No.:
    MSPP-CA4505
  • Supplier no.:
    CA4505
Specifications
About this item
Fits GL45 thread safety caps, GL38 bottle and Canary-Safe™ caps.
Fits to most common 4 L solvent bottles.