- Antibody Type:Secondary
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western Blot:Yes
- Size:100 µg
- Shipping Temperature:Ambient
- Purification:immunoaffinity chromatography
- Cat. No.:76225-984
Anti-Mouse IgG DyLight549 Antibody generated in rabbit detects reactivity to Mouse IgG1, IgG2a, IgG2b and IgG3. Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The whole IgG molecule possesses both the F(c) region, recognized by high-affinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site. IgG1, IgG2a, IgG2b and IgG3 chains of the antibody molecule are present. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition.
Mouse IgG subclass pan reactive Secondary Antibody is designed for STED microscopy, FRET, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. Rabbit anti-mouse IgG antibody DyLight™ 549 is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation. Mouse IgG secondary antibody is available in a variety of formats.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DL549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse