Specifications
- Antibody Type:Primary
- Antigen Symbol:ZC3H12C
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75933-702
- Supplier no.:600-401-GC6
Specifications
About this item
ZC3H12C, also known as MCPIP3, is a member of a family of novel CCCH-zinc finger proteins that includes ZC3H12A, a protein that is thought to be involved in macrophage activation, host immunity and inflammatory diseases. Similar to ZC3H12A, ZC3H12C expression in macrophages is highly increased after treatment with lipopolysaccharide (LPS), suggesting it also may play a role in host immunity and inflammatory response.
Anti-ZC3H12C Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 99 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: ZC3H12C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: