Specifications
- Antibody Type:Primary
- Antigen Name:VDAC1
- Antigen Symbol:VDAC1
- Clonality:Monoclonal
- Clone:S152B-23
- Conjugation:RPE (R-Phycoerythrin)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Western Blot:Yes
- Size:100 µg
- Shipping Temperature:Wet Ice
- Purification:Protein G purified
- Cat. No.:75933-544
- Supplier no.:200-308-W70
Specifications
About this item
Voltage-dependent anion-selective channel protein 1 (also known as VDAC, VDAC1 or outer mitochondrial membrane protein porin 1) is the the outer mitochondrial membrane receptor for hexokinase and BCL2L1. VDAC forms a channel through the mitochondrial membrane and is involved in small molecule diffusion, cell volume regulation and apoptosis. VDAC may participate in the formation of the permeability transition pore complex (PTPC), which is responsible for the release of mitochondrial products that triggers apoptosis. Anti-VDAC1 is ideal for research in Cell Biology and Apoptosis.
Anti-VDAC1 R-PE Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~30kDa on specific lysates or tissues. Does not cross react with VDAC2 or VDAC3 (based on KO validation results). Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: VDAC1
Clonality: Monoclonal
Clone: S152B-23
Conjugation: RPE
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: