- Antibody Type:Primary
- Antigen Symbol:UNG
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75933-446
- Supplier no.:600-401-FV0
The human uracil-DNA glycosylase (UNG) gene encodes both mitochondrial (UNG1) and nuclear (UNG2) forms through differentially regulated promotes and alternative splicing. While UNG2 is the major enzyme in the base excision repair pathway that removes uracil residues from nuclear DNA that arise through either misincorporation during replication or cytosine deamination, inhibition of UNG1 by uracil glycosylase inhibitor did not lead to increased levels of spontaneous or induced mitochondrial DNA mutations. However, decreased levels of UNG activity and increased oxidative damage to mitochondrial DNA were seen in older mice, suggesting that mitochondrial DNA repair mechanisms may be involved in various neurodegenerative disorders in an age-dependent manner. This UNG1 antibody will not cross-react with UNG2.
Anti-UNG1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 34 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: UNG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: