Specifications
- Antibody Type:Primary
- Antigen Symbol:TXNIP
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75933-288
- Supplier no.:600-401-FT0
Specifications
About this item
Thioredoxin-interacting protein (TXNIP) belongs to the arrestin family and plays a critical role in the antioxidant defense mechanisms of hematopoietic cells by activating the p53 pathway during oxidative stress (1,2). It functions as a transcriptional repressor and acts as an oxidative stress mediator by inhibiting thioredoxin activity (2). TXNIP expression is reduced in many types of tumors, and TXNIP overexpression inhibits tumor growth by blocking cell-cycle progression (3). It has recently reported that TXNIP deficiency correlates with a high incidence of hepatocellular carcinoma (HCC) (4). TXNIP and p53 interactions could potentially be a therapeutic target for oxidative stress-related diseases such as hematopoietic malignancies and metabolic diseases (5).
Anti-TXNIP Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 44 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: TXNIP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: