Specifications
- Antibody Type:Primary
- Antigen Symbol:SUC1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Western Blot:Yes
- Size:100 µg
- Shipping Temperature:Ambient
- Purification:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography
- Cat. No.:75929-184
- Supplier no.:200-1061890100
Specifications
About this item
Anti-Invertase antibody recognizes invertase which hydrolyzes the alpha-1,4-glycosidic bond between alpha-D-glucose and beta-D-fructose of sucrose. Bees use invertase to convert nectar to honey. In the confectionary industry, yeast-derived invertase is used to hydrolyze sucrose to glucose and fructose as fructose is more sweeter and less prone to crystallization.
Anti-Invertase antibody is ideal for investigators interested in Metabolism and Cell Biology.
Anti-Invertase antibody has been assayed against 1.0 ug of Invertase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature. A working dilution of 1:4,000 to 1:20,000 of the reconstitution concentration is suggested for this product.
Type: Primary
Antigen: SUC1
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity: