Specifications
- Antibody Type:Primary
- Antigen Symbol:SPP1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75934-168
- Supplier no.:600-401-ET6
Specifications
About this item
The secreted protein 1 (SPP1), also known as osteopontin, is a major noncollagenous protein of bone, but is also found in the extracellular matrix of other mineralized tissues and in bodily fluids. In bone, SPP1 is produced by osteoblasts, osteocytes, macrophages, and osteoclasts (1,2). SPP1 binds to cells through integrin and non-integrin receptors, as well as the adhesion receptor CD44 in an RGD-independent manner, enabling SPP1 to induce a number of functional effects including macrophage chemotaxis, cytoprotection, and regulation of T helper type 1 cells (2). SPP1 can regulate biomineralization by inhibiting the formation of hydroxyapatite (3) and the growth of calcium oxalate crystals (4).
Anti-SPP1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 35 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: SPP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: